Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RB Mouse anti-Human, Clone: MM0374-5P14, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2116730.025MG
Description
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17RB | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q9NRM6 | |
IL17RB | |
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-17B R extracellular domain | |
0.025 mg | |
Cytokine Research | |
55540 | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
MM0374-5P14 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only