Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RD/SEF Antibody (309511), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB22751MFV500
Description
IL-17RD/SEF Monoclonal antibody specifically detects IL-17RD/SEF in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| IL-17RD/SEF | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54756 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 309511 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17RD, Ala27-Arg299, Accession # Q8NFM7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction