Learn More
MBL International Corporation Human IL-18 ELISA Kit
SDP
Supplier:  MBL International Corporation 7620
Encompass_Preferred
Size:96 Wells; The Human Il-18 ELISA Kit is based on sandwich ELISA and is capable of measuring human Il-18. This kit has a high sensitivity (12.5 pg/ml) and continues to be gather multiple citations every year.With regard to diseases, high levels of IL-18 are detected in the blood of patients with allergic diseases (bronchial asthma, atopic dermatitis, allergic rhinitis) and those with autoimmune diseases (rheumatoid arthritis, systemic lupus erythematosus [SLE], adult Stills disease, multiple sclerosis, Chrohns disease, ulcerative colitis). IL-18 is also found in the urine of patients with acute renal disorder and appears to mediate the progression of type 2 diabetes.Human IL-18 ELISA Kit
Catalog No. 50-446-36
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.