Learn More
IL-18 R beta/IL-1 R7/ACPL Rabbit anti-Human, Alexa Fluor 594, Clone: 122, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP289316AF594
Catalog No. NB066216
Add to cart
Description
Description
IL-18 R beta/IL-1 R7/ACPL Monoclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
Specifications
IL-18 R beta/IL-1 R7/ACPL
122
Sandwich ELISA Detection
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor 594
50mM Sodium Borate
Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 R beta/IL-1 R7/ACPL (NP_003844.1; Met1-Gly357).
Protein A purified
RUO
Primary
8807