Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-19 Antibody Pair [HRP]
SDP

Polyclonal/Monoclonal Antibody Pair

$832.00

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009855
SDP
View Documents
Novus Biologicals™
NBP27940815PLATES
15 Plates
Each of 1 for $832.00
Add to Cart
 
Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human IL-19.
Specifications

Specifications

31.25 pg/mL
Sandwich ELISA
Antibody Pairs
Human
IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19
IL19
  • Standard: Each vial contains 30 ng of recombinant Human IL-19.
  • Capture Antibody: 1 mg/mL of mouse anti-Human IL-19 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human IL-19 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Biologically Active Proteins, Cytokine Research
    IL-19
    31.25-2000 pg/mL
    HRP
    This solid phase sandwich ELISA utilizes an antibody specific for Human IL-19.
    Sandwich ELISA
    29949
    Rabbit/Mouse
    RUO
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    SDS
    Documents

    Documents

    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit