Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-19 Monoclonal Antibody (C9)
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | IL-19 |
---|---|
Clone | C9 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541618
|
Invitrogen™
MA541618 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543694
|
Invitrogen™
MA543694 |
20 μL |
Each for $243.00
|
|
|||||
Description
IL-19 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.Specifications
IL-19 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UHD0 | |
29949 | |
Recombinant protein Interleukin 19. The antigen corresponds to amino acid range 25-177 of the target protein. | |
Antibody |
C9 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
IL19 | |
il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1 | |
IL19 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title