Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1ra/IL-1F3/IL1RN Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271022AF405

Catalog No. N271022A405


Add to cart

Description

Description

IL-1ra/IL-1F3/IL1RN Monoclonal antibody specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

IL-1ra/IL-1F3/IL1RN
OTI2B1
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
IL1RN
Full length recombinant protein of human IL-1ra/IL-1F3/IL1RN (NP_776215) produced in E.coli.
Immunogen affinity purified
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3557.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit