Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | IL-1ra/IL-1F3/IL1RN |
---|---|
Clone | 1 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for ELISA.Specifications
IL-1ra/IL-1F3/IL1RN | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
Recombinant Human IL-1ra/IL-1F3/IL1RN protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
0.2 um filtered solution in PBS | |
3557 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title