Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1Rrp2/IL-1R6, Mouse anti-Human, DyLight 488, Clone: 15G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233299G

Catalog No. NB005782

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

IL-1Rrp2/IL-1R6 Monoclonal antibody specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-1Rrp2/IL-1R6
15G9
Western Blot
Purified, Conjugated
IL1RL2
Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Protein G purified
RUO
Primary
Human
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit