Learn More
Novus Biologicals™ IL-2 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192661
Catalog No. NBP192661
Add to cart
Description
Description
The Mouse IL-2 ELISA Kit (Colorimetric) measures mouse IL-2 in cell culture supernatant, serum.
Specifications
Specifications
15.6 - 1,000pg/mL
3558
ELISA Kit
5.3pg/mL
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity
aldesleukin, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-2, Biotin-Conjugate anti-mouse IL-2 monoclonal antibody, Streptavidin-HRP, Mouse IL-2 Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
IL-2
IL2