Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2, Mouse anti-Human, Clone: 5334R, R&D Systems™

Mouse Monoclonal Antibody

$155.80 - $372.00

Specifications

Antigen IL-2
Clone 5334R
Immunogen E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB202R100
View Documents
R&D Systems
MAB202R100
100 ug
Each for $372.00
Add to cart
 
MAB202RSP
View Documents
R&D Systems
MAB202RSP
25 ug
Each for $155.80
Add to cart
 
Description

Description

IL-2 Monoclonal antibody specifically detects IL-2 in Human Samples. It is validated for Neutralization, Immunocytochemistry.
Specifications

Specifications

IL-2
E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577
Primary
Human
Unconjugated
3558
Purified
Neutralization, Immunocytochemistry
RUO
5334R
Mouse
Monoclonal
IgG1
aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit