Learn More
Invitrogen™ IL-2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA546923
Description
In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human IL-2, recombinant mouse IL-2, recombinant rat IL-2, recombinant feline IL-2, recombinant canine IL-2, recombinant equine IL-2, recombinant cotton rat IL-2, and recombinant porcine IL-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
IL-2 | |
Polyclonal | |
Unconjugated | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
280822 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P05016 | |
IL2 | |
E. coli-derived recombinant bovine IL-2 Ala21-Thr155. | |
100 μg | |
Primary | |
Bovine | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.