Learn More
IL-2 Rat anti-Human, Alexa Fluor® 488, Clone: MQ1-17H12, Invitrogen™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Manufacturer:  Invitrogen RHCIL220
Catalog No. 50-113-7574
Add to cart
Description
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
Specifications
IL-2
Monoclonal
Alexa Fluor® 488
IL2
Liquid
IL2
Human IL-2.
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MQ1-17H12
PBS with BSA and 0.1% sodium azide
P60568
T-cell growth factor, TCGF, interleukin-2
Rat
IgG2a, kappa
500 μL
4° C
Primary
3558