Learn More
IL-2 Monoclonal Antibody (MQ1-17H12), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$335.65
Specifications
Antigen | IL-2 |
---|---|
Clone | MQ1-17H12 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9047
|
Invitrogen
48702942 |
100 Tests |
Each of 1
for $335.65
Save 5% Reg : |
|
|||||
Description
Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody. Applications Reported: This MQ1-17H12 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MQ1-17H12 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry.Specifications
IL-2 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P60568 | |
IL2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2 |
MQ1-17H12 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Human | |
3558 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.