Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Rat anti-Mouse, BV421, Clone: JES6-5H4, BD Horizon™

Rat Monoclonal Antibody

Supplier:  BD Biosciences 566296

Catalog No. BDB566296


Add to Cart

Description

Description

IL-2 Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-2
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
50 μg
Cell Biology
Mouse
Purified
Flow Cytometry, Flow Cytometry (Intracellular Staining)
JES6-5H4
Brilliant Violet 421
Il2; Interleukin-2; T-cell growth factor; TCGF
Mouse IL-2 Recombinant Protein
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
IgG2b
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.