Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-20 Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794075PLATES

 View more versions of this product

Catalog No. NB009854


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-20.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
6.25 pg/mL
50604
RUO
IL-20
Biologically Active Proteins, Signal Transduction
Human
IL20
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-20.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human IL-20 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Human IL-20 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 16 ng of recombinant Human IL-20.
  • Storage is content dependent.
    Mouse/Rabbit
    5 Plates
    6.25-400 pg/mL
    Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit