Learn More
IL-20 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Supplier: Pepro Tech Inc 500P190G50UG

Description
AA Sequence of recombinant protein: MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-20. Anti-Human IL-20-specific antibody was purified by affinity chromatography employing an immobilized Human IL-20 matrix. Sandwich ELISA: To detect Human IL-20 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-20 (500-P190GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-20. Western Blot: To detect Human IL-20 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-20 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL-20 Polyclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISA, Western BlotSpecifications
IL-20 | |
Polyclonal | |
PBS with no preservative | |
Q9NYY1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
50604 | |
-20°C |
ELISA, Western Blot | |
Unconjugated | |
Il20 | |
cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10 | |
E.coli-derived Recombinant Human IL-20 | |
50 μg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.