Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 R beta/FNDC6 Rat anti-Human, Mouse, DyLight™ 550, Clone: 20RNTC, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200466R
Description
IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-20 R beta/FNDC6 | |
Monoclonal | |
DyLight 550 | |
50 mM sodium borate with 0.05% sodium azide | |
IL20RB | |
The immunogen for this antibody is FNDC6 (IL-20R2). | |
0.1 mL | |
53833.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
20RNTC | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
Rat | |
Protein A or G purified | |
Primary | |
Human, Mouse | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title