Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

IL-20 Rabbit anti-Human, Alexa Fluor 647, Clone: 17, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290161AF647

Catalog No. NB070490



Description

Description

IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISA
Specifications

Specifications

IL-20
17
ELISA
Purified
Rabbit
IgG
0.1 mL
Biologically Active Proteins, Signal Transduction
Primary
50604
ELISA
Alexa Fluor 647
50mM Sodium Borate
Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Recombinant Human IL-20 Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit