Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Antibody (227), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326073MFV450
Description
IL-21 Monoclonal antibody specifically detects IL-21 in Mouse samples. It is validated for ELISASpecifications
IL-21 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
227 | |
ELISA | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Rabbit immunized with purified, recombinant Mouse IL-21 (Uniprot#: Q9ES17, Pro25-Ser146) | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
59067 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction