Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Mouse anti-Human, Alexa Fluor™ 532, Clone: 14k5A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227337AF532

Catalog No. NP227337A32


Add to cart

Description

Description

IL-21 Monoclonal antibody specifically detects IL-21 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-21
14k5A9
Western Blot
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
IL21
Full length recombinant protein
0.1 mL
Primary
59067
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit