Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Mouse anti-Human, Alexa Fluor™ 750, Clone: 14k5H3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227336AF750
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IL-21 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
IL21 | |
Full length recombinant protein | |
Primary | |
Human | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
14k5H3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Mouse | |
0.1 mL | |
59067 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title