Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Mouse anti-Human, Clone: 148002, R&D Systems™
Mouse Monoclonal Antibody
$136.67 - $415.56
Specifications
Antigen | IL-21 |
---|---|
Clone | 148002 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1500100
|
R&D Systems
MAB1500100 |
100 μg |
Each for $415.56
|
|
MAB1500SP
|
R&D Systems
MAB1500SP |
25 μg |
Each for $136.67
|
|
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for ELISA.Specifications
IL-21 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9HBE4 | |
59067 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
148002 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
E. coli-derived recombinant human IL-21, Gln25-Ser155, Accession # Q9HBE4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title