Learn More
IL-21R Mouse anti-Human, Alexa Fluor 405, Clone: 152512, R&D Systems
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB9911V100UG
Catalog No. FAB9911V100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-21R Monoclonal antibody specifically detects IL-21R in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-21R
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9HBE5
IL21R
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R Cys20-Pro236 Accession # Q9HBE5
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human IL-21 R in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh)IL-2 R beta or rmIL-21 R.
Flow Cytometry
152512
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Mouse
IgG1
RUO
Primary
50615
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)