Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ IL-22 Antibody Pair [HRP]
Monoclonal Antibody Pair
$832.00
Specifications
Assay Range | 2.03 pg/mL |
---|---|
Assay Sensitivity | 2.03-130 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009851
|
Novus Biologicals™
NBP27940615PLATES |
15 Plates |
Each of 1 for $832.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-22.Specifications
2.03 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-21, IL-22IL21, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 21, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
IL22 | |
RUO | |
Storage is content dependent. |
2.03-130 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-22. | |
Sandwich ELISA | |
50616 | |
Rabbit/Mouse | |
Biologically Active Proteins, Cytokine Research, Innate Immunity | |
IL-22 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title