Learn More
IL-22 Mouse anti-Human, Alexa Fluor™ 350, Clone: 8F7H11.2B9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227320AF350
Catalog No. NP227320A30
Add to cart
Description
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-22
8F7H11.2B9
Western Blot
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-21, IL-22IL21, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 21, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
IL22
Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
0.1 mL
Primary
50616