Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Mouse anti-Human, Alexa Fluor™ 750, Clone: 8F7H11.2B9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227320AF750

Catalog No. NP227320A70


Add to cart

Description

Description

IL-22 Monoclonal antibody specifically detects IL-22 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-22
8F7H11.2B9
Western Blot
Conjugated, Purified
IL22
Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Protein G purified
Biologically Active Proteins, Cytokine Research, Innate Immunity
Primary
50616.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit