Learn More
IL-22 Monoclonal Antibody (22URTI), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$393.65
Specifications
Antigen | IL-22 |
---|---|
Clone | 22URTI |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-161-06
|
Invitrogen
46722942 |
100 Tests |
Each of 1
for $393.65
Save 12% Reg : |
|
|||||
Description
Description: The 22URTI monoclonal antibody reacts with human interleukin(IL)-22. IL-22 is a 20 kDa member of the IL-10 cytokine family that is secreted primarily by Th17 and NK cells. Nevertheless, other T cells have also been shown to produce IL-22. In in vitro Th17 cultures, induction of IL-22 expression is greater in response to IL-23 than IL-6 or TGF beta, suggesting that this cytokine may be secreted by more fully differentiated Th17 cells in vivo. A heterodimer consisting of IL-10R2 and IL-22R1 serves as the receptor for IL-22. Applications Reported: This 22URTI antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 22URTI antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of restimulated, Th17-polarized CD4+ human peripheral blood mononuclear cells (normal human peripheral blood cells) or restimulated total normal human peripheral blood cells. This can be used at 5 μL (0.015 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm).
IL-22 Monoclonal antibody specifically detects IL-22 in Human samples. It is validated for Flow Cytometry.Specifications
IL-22 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9GZX6 | |
IL22 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL22 |
22URTI | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
50616 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.