Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22 Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB100737
Description
IL-22 Polyclonal specifically detects IL-22 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-22 | |
Polyclonal | |
Unconjugated | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
IL22 | |
Synthetic peptide: CKLEVSNFQQPYIVNRTFMLAKEASLAD, corresponding to amino acids 40-67 of Mouse IL22 precursor. | |
0.1 mg | |
Innate Immunity | |
50616 | |
Human, Mouse | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
ELISA 1:100000, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500 | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
The peptide sequence is <50% identical to other interleukins in this region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title