Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22R alpha 1 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
$177.31
Specifications
Antigen | IL-22R alpha 1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017810
|
Novus Biologicals
NB1007400.025MG |
0.025 mg |
Each for $177.31
|
|
NB100740
|
Novus Biologicals
NB100740 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-22R alpha 1 | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Goat | |
Human | |
CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
IL22RA1 | |
IgG | |
Affinity Purified | |
IL22 Receptor |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry | |
Unconjugated | |
RUO | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
58985 | |
Synthetic peptide: DSGPEGTPDTVYSIEYKTYGERDWVAKK, corresponding to N terminal amino acids 42-70 of Human IL22 Receptor. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title