Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-23 Antibody (207), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326067UV
Description
IL-23 Monoclonal antibody specifically detects IL-23 in Mouse samples. It is validated for ELISASpecifications
IL-23 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
207 | |
ELISA | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12B, IL-23, IL-23 subunit alpha, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 12, p40, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-12 beta chain, interleukin-12 subunit beta, Interleukin-23 subunit p19, interleukin-six, G-CSF related factor, JKA3 induced upon T-cell activation, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, P19, SGRF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-23 (Uniprot#: Q9EQ14 and P43432, Met1-Ala196 and Met1-Ser335) | |
0.1 mL | |
Cancer, Cytokine Research, Innate Immunity | |
51561 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction