Learn More
IL-23 p19 Rat anti-Mouse, Alexa Fluor 647, Clone: N71-1183, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565317
Catalog No. BDB565317
Add to cart
Description
Description
IL-23 p19 Monoclonal antibody specifically detects IL-23 p19 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
IL-23 p19
N71-1183
Alexa Fluor 647
Purified
Rat
Rat IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
interleukin-23 subunit alpha; IL-23-A; IL-23p19; Il23a
Mouse IL-23p19 Recombinant Protein
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.