Learn More
IL-23 p19 Rat anti-Mouse, Clone: N71-1183, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565344
Catalog No. BDB565344
Add to cart
Description
Description
IL-23 p19 Monoclonal antibody specifically detects IL-23 p19 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
IL-23 p19
N71-1183
Unconjugated
Purified
Rat
Rat IgG1, κ
RUO
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
1.0mg/mL
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2µm sterile filtered. Endotoxin level is ≤0.01 EU/µg (≤0.001 ng/µg) of protein as determined by the LAL assay.
interleukin-23 subunit alpha; IL-23-A; IL-23p19; Il23a
Mouse IL-23p19 Recombinant Protein
0.5mg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.