Learn More
IL-23 R Rat anti-Mouse, PerCP, Clone: 753317, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB16861C100UG
 View more versions of this product
Catalog No. FB16861C100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-23R
753317
Flow Cytometry 0.25-1 μg/106 cells
Q8K4B4
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Rat
IgG1
100 μg
Primary
149233.0
Mouse
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant, Conjugated
IL23R
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession No. Q8K4B4
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects endogenous mouse IL-23 R by flow cytometry.