Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-23R PerCP-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB16861C025UG
This item is not returnable.
View return policy
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-23R | |
Monoclonal | |
PerCP | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects endogenous mouse IL-23 R by flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
753317 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q8K4B4 | |
IL23R | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession No. Q8K4B4 | |
25 μg | |
149233.0 | |
Mouse | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction