Learn More
IL-23 Receptor Rat anti-Mouse, BB515, Clone: O78-1208, BD
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566212
Catalog No. BDB566212
Add to cart
Description
Description
The O78-1208 monoclonal antibody specifically binds to the mouse Interleukin-23 Receptor (IL-23R) subunit that is encoded by the il23r /gene. The IL-23R subunit is a type I transmembrane glycoprotein and member of the hemopoietin receptor superfamily. The mouse IL-23 Receptor complex is comprised of IL-23R and IL-12 receptor beta 1 (IL-12Rβ1) subunits. The IL-23R complex can bind IL-23, a cytokine that plays roles in innate and adaptive immunity as well as in autoimmune diseases, eg, by the generation and maintenance of Th17 cells. Mouse IL-23R is expressed by activated/memory CD4+ T cells, Th1, Th2 and Th17 cells,γδ T cells,dendritic cells and macrophages as determined by IL-23R mRNA expression and IL-23R-GFP reporter mouse studies. The IL-23-bound IL-23R complex transduces an intracellular signal pathway mediated by a Jak-STAT signaling cascade.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490 nm and an emission max of 515 nm, BD Horizon BB515 can be excited by the 488 nm laser and detected in a standard FITC set (eg, 530/30nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector. BB515 is a dye that was exclusively developed by BD Biosciences as a brighter alternative to FITC. This dye is up to seven times brighter than FITC and has less spillover into the PE channel. Due to similar excitation and emission properties, BB515, FITC, and Alexa Fluor™ 488 cannot be used simultaneously.
Specifications
Specifications
IL-23 Receptor
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Il23r, IL-23R, Interleukin 23 receptor
Mouse IL-23 Receptor Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
RUO
Primary
Murine
O78-1208
BB515
Affinity Purified
Rat
Rat LOU, also known as Louvain, LOU/C, LOU/M IgG1, κ
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal