Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-25R (IL-17RB) Rat anti-Mouse, PE, Clone: MUNC33, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 12736180

Encompass

 View more versions of this product

Catalog No. 50-112-8994


Add to cart

Description

Description

Description: This MUNC33 monoclonal antibody reacts with mouse interleukin-25 receptor (IL-25R), also known as IL-17RB. IL-25R is a 55 kDa transmembrane protein that dimerizes with IL-17RA to form the functional receptor for IL-25 (IL-17E). IL-25R is detectable in various tissues, including the lung and gastrointestinal tract and exists as both membrane bound and soluble form. IL-25/IL-25R interaction has been shown to activate the NF-kB, MAPK, and JNK pathways as well as gene transcription by STAT6, GATA-3, and NFATc1. IL-25R signals in T helper 2 (Th2) cells, type 2 innate lymphoid (ILC2) cells and some iNKT cells to promote differentiation and effector function. Whereas IL-25-deficient mice have an impaired Th2 response to parasitic helminth infection, elevated expression of IL-25 and IL-25R is observed in patients with chronic asthma and atopic dermatitis. Applications Reported: This MUNC33 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MUNC33 antibody has been tested by flow cytometirc analysis of mouse lymph node cells or IL-25R-transfected cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are IL-17B, IL-17C, and IL-17E. Like IL-17, IL-17B, C, and E are able to induce proinflammatory responses but don′t bind to the IL-17 receptor suggesting additional IL-17R related receptors might exist. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in kidney and liver and at moderate level in small intestine, colon, and brain.
Specifications

Specifications

IL-25R (IL-17RB)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9JIP3
Il17rb
Affinity chromatography
RUO
50905
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MUNC33
PE
Il17rb
IL-17B, IL-20, MGC138900, MGC138901, ZCYTO7
Rat
25 μg
Primary
Mouse
Antibody
IgG2a κ
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.