Learn More
IL-26/AK155 Mouse anti-Human, Alexa Fluor 700, Clone: MM0399-6D20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211703AF700
Catalog No. NB211703700
Description
Description
IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-26/AK155
MM0399-6D20
Western Blot
Purified
IL26
Human IL26
Protein G purified
Cytokine Research
Primary
55801
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
AK155IL-26interleukin-26, interleukin 26, Protein AK155
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human