Learn More
IL-27 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 241651AP
Catalog No. 50-173-4249
Add to cart
Description
Description
The protein encoded by IL-27 is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR).
Specifications
Specifications
IL-27
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NEV9
IL 27, IL 27 A, IL 27 subunit alpha, IL 27A, IL27, IL27 A, IL27A, IL27p28, IL30, interleukin 27, Interleukin 27 subunit alpha, p28
Rabbit
IgG
150 μL
-20°C
Primary
246778
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IL27
Liquid
IL27
IL-27 Fusion Protein Ag21436
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human