Learn More
IL-28A/IFN-lambda 2 Mouse anti-Human, Alexa Fluor 700, Clone: MM0400-6Z37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211705AF700
Catalog No. NB211705700
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IL-28A/IFN-lambda 2 Monoclonal antibody specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-28A/IFN-lambda 2
MM0400-6Z37
Western Blot
Purified
IFNL2
Human IL28a
Protein G purified
Biologically Active Proteins
Primary
282616
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
interleukin-28A, IL28A, IL-28A, interferon, lambda 2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human