Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-28A/IFN-lambda 2 Mouse anti-Human, Clone: MM0400-6Z37, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | IL-28A/IFN-lambda 2 |
---|---|
Clone | MM0400-6Z37 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP211705
|
Novus Biologicals
NBP211705 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018862
|
Novus Biologicals
NBP2117050.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
IL-28A/IFN-lambda 2 Monoclonal antibody specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Western Blot.Specifications
IL-28A/IFN-lambda 2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
interleukin-28A, IL28A, IL-28A, interferon, lambda 2 | |
IFNL2 | |
IgG1 | |
Protein G purified |
MM0400-6Z37 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Q8IZJ0 | |
282616 | |
Human IL28a | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title