Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28B Mouse anti-Human, Clone: GT727, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527867

Catalog No. PIMA527867


Add to cart

Description

Description

Positive Control: GFP-human IL28-transfected 293T cells Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
Specifications

Specifications

IL-28B
Monoclonal
1 mg/ml
PBS with no preservative
Q8IZI9
IFNL3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human IL28B. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Western Blot
GT727
Unconjugated
IFNL3
Liquid
Mouse
IgG2b
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
282617
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit