Learn More
IL-29 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179951AP
Catalog No. 50-173-1924
Add to cart
Description
Description
IL29 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28A (IL28A), and interleukin 28B (IL28B) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
Specifications
Specifications
IL-29
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IU54
Cytokine Zcyto21, IFN lambda 1, IFNL1, IL 29, IL29, IL-29, Interferon lambda 1, Interleukin 29, ZCYTO21
Rabbit
IgG
150 μL
-20°C
Primary
282618
Western Blot
Unconjugated
IFNL1
Liquid
IFNL1
IL-29 Fusion Protein Ag12444
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human