Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 R beta Antibody (TM-b1), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143355JF549
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-2Rbeta | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL2RB | |
The immunogen for this antibody was Purified CD122. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
TM-b1 | |
Flow Cytometry | |
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
Rat | |
Protein A or G purified | |
Immunology | |
3560 | |
Store at 4°C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction