Learn More
IL-3 beta Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Supplier: Pepro Tech Inc 500P177G50UG

Description
AA Sequence of recombinant protein: MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC. Preparation: Produced from sera of goats immunized with highly pure Recombinant Rat IL-3beta. Anti-Rat IL-3beta-specific antibody was purified by affinity chromatography employing an immobilized Rat IL-3beta matrix. Sandwich ELISA: To detect Rat IL-3beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat IL-3beta (500-P177GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-3beta. Western Blot: To detect Rat IL-3beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-3beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL-3 beta Polyclonal antibody specifically detects IL-3 beta in Rat samples. It is validated for ELISA, Western BlotSpecifications
IL-3 beta | |
Polyclonal | |
PBS with no preservative | |
P04823 | |
E.coli-derived Recombinant Rat IL-3beta | |
50 μg | |
Primary | |
Rat | |
Lyophilized |
ELISA, Western Blot | |
Unconjugated | |
Il3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
24495 | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.