Learn More
IL-3 R alpha/CD123 Mouse anti-Human, DyLight 405, Clone: 8E11C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237354V
Catalog No. NBP237354V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IL-3 R alpha/CD123
8E11C5
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
IL3RA
Purified recombinant fragment of human IL3RA (AA: 200-305) expressed in E. Coli.
Protein A or G purified
B Cell Development and Differentiation Markers, Cytokine Research, Phospho Specific
Primary
3563
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human