Learn More
IL-3R alpha/CD123 Rat anti-Mouse, Alexa Fluor 750, Clone: 151231, R&D Systems
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB983S100UG
Catalog No. FAB983S100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-3R alpha/CD123
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P26952
IL3RA
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-3 R alpha/CD123 Ser17-Lys331 Accession # P26952
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-3 R alpha and recombinant mouse IL-3 R beta is observed.
Flow Cytometry
151231
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Rat
IgG2a
RUO
Primary
3563
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)