Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-31 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176419MFV610
Description
IL-31 Polyclonal antibody specifically detects IL-31 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
IL-31 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
IL-31interleukin-31, interleukin 31 | |
Antibody was raised against a 18 amino acid synthetic peptide from near the center of human IL-31. The immunogen is located within amino acids 60 - 110 of IL-31. Amino Acid Squence: CLSPDAQPPNNIHSPAIR | |
0.1 mL | |
Biologically Active Proteins | |
386653 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction