Learn More
IL-33, Mouse anti-Human, Clone: 6H496, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP280809
Catalog No. NB013882
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human Samples. It is validated for Western Blot, Simple Western.
Specifications
Specifications
IL-33
Monoclonal
0.5 mg/mL
Western Blot 1-10 μg/mL, Simple Western : 1:50
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot
6H496
Unconjugated
PBS
IL33
A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
0.1 mg
Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
90865
Store at 4°C. Do not freeze.
IgG1 κ