Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-33 Mouse anti-Human, Clone: IL33305B, Novus Biologicals™
Mouse Monoclonal Antibody
$350.00 - $542.50
Specifications
Antigen | IL-33 |
---|---|
Clone | IL33305B |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:2000-1:5000, ELISA 1:2000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200, Direct ELISA 1:2000-1:5000, Block/Neutralize |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA, Neutralization |
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Direct ELISA, Block/Neutralize.Specifications
IL-33 | |
1 mg/ml | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA, Neutralization | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
0.2 um-filtered solution in PBS, pH 7.4 | |
90865 | |
IgG2a κ | |
Protein G purified |
IL33305B | |
Western Blot 1:2000-1:5000, ELISA 1:2000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200, Direct ELISA 1:2000-1:5000, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
Recombinant human IL-33. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title